Invitrogen
Manufacturer Code:PA5102514
Catalog # PIPA5102514
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human IL1R2 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: antigen CDw121b; antigen CDw121b CD121 antigen-like family member B CD121b CDw121b IL-1 receptor beta chain IL-1 type II receptor Il1r2 Il-1r2 IL-1R-2 Il1r-2 IL1R2c IL1RB IL-1R-beta IL-1RT2 IL-1RT-2 ILR2 interleukin 1 receptor type 2 interleukin 1 receptor type II variant 3 interleukin 1 receptor type II interleukin-1 receptor beta Interleukin-1 receptor type 2 Interleukin-1 receptor type 2 membrane form Interleukin-1 receptor type 2 soluble form Interleukin-1 receptor type II mIL-1R2 sIL-1R2 type II interleukin-1 receptor beta; CD121 antigen-like family member B; CD121b; CDw121b; IL-1 receptor beta chain; IL-1 type II receptor; IL-1R-2; IL-1R-beta; IL-1RT-2; IL-1RT2; interleukin 1 receptor type II variant 3; interleukin 1 receptor, type II; Interleukin-1 receptor beta; Interleukin-1 receptor type 2; Interleukin-1 receptor type 2, membrane form; Interleukin-1 receptor type 2, soluble form; Interleukin-1 receptor type II; mIL-1R2; sIL-1R2; type II interleukin-1 receptor, beta
Gene Aliases: CD121b; CDw121b; IL-1R-2; Il-1r2; IL-1RT-2; IL-1RT2; Il1r-2; IL1R2; IL1R2c; IL1RB
UniProt ID: (Human) P27930, (Mouse) P27931
Entrez Gene ID: (Human) 7850, (Mouse) 16178
Molecular Function: cytokine receptor receptor type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.