Invitrogen
Manufacturer Code:PA546930
Catalog # PIPA546930
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Sf 21-derived recombit human IL-1 RI |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CD121 antigen-like family member A; CD121a; CD121A D2S1473 IL-1R-alpha IL1R IL1RA P80 CD121 antigen-like family member A IL-1R-1 IL-1RT-1 IL-1RT1 antigen CD121a interleukin 1 receptor alpha type I interleukin receptor 1 interleukin-1 receptor alpha interleukin-1 receptor type 1 interleukin-1 receptor type I; IL-1R-1; IL-1R-alpha; IL-1RT-1; IL-1RT1; interleukin 1 receptor alpha, type I; interleukin 1 receptor, type I; Interleukin-1 receptor alpha; Interleukin-1 receptor type 1; Interleukin-1 receptor type 1, membrane form; Interleukin-1 receptor type 1, soluble form; Interleukin-1 receptor type I; mIL-1R1; p80; sIL-1R1
Gene Aliases: CD121A; D2S1473; IL-1R-alpha; IL1R; IL1R1; IL1RA; IL1RT1; P80
UniProt ID: (Human) P14778
Entrez Gene ID: (Human) 3554
Molecular Function: cytokine receptor receptor type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.