Abnova
Manufacturer Code:MAB3004
Catalog # 89-111-197
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6E10 |
Immunogen | Recombinant protein corresponding to human IL1B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Mouse monoclonal antibody raised against recombinant IL1B.
For Research Use Only
Protein Aliases: Catabolin; IL-1; IL-1 beta; IL1-BETA; IL1F2; interleukin 1, beta; Interleukin-1 beta; preinterleukin 1 beta; pro-interleukin-1-beta
Gene Aliases: IL-1; IL1-BETA; IL1B; IL1F2
UniProt ID: (Human) P01584
Entrez Gene ID: (Human) 3553
Molecular Function: cytokine interleukin superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.