OriGene
Manufacturer Code:CF506850
Catalog # 50-166-4481
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2F11 |
Immunogen | Full length human recombit protein of human IL1A produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Hematopoietin-1; IL-1 alpha; IL-1A IL1 IL1-ALPHA IL1F1; interleukin 1, alpha; Interleukin-1 alpha; preinterleukin 1 alpha; pro-interleukin-1-alpha
Gene Aliases: IL-1A; IL1; IL1-ALPHA; IL1A; IL1F1
UniProt ID: (Human) P01583
Entrez Gene ID: (Human) 3552
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.