Invitrogen
Manufacturer Code:PA575838
Catalog # PIPA575838
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human IL18RAP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Accessory protein-like; AcPL; AcPL accessory protein-like); CD218 antigen-like family member B; CD218b; CDw218b; cluster of differentiation w218b; IL-18 receptor accessory protein; IL-18 receptor beta; IL-18R-beta; IL-1R accessory protein-like; IL-1R-7; IL-1RAcPL; Interleukin-1 receptor 7; Interleukin-18 receptor accessory protein; Interleukin-18 receptor accessory protein IL-18 receptor accessory protein IL-18RAcP Accessory protein-like AcPL CD218 antigen-like family member B CDw218b IL-1R accessory protein-like IL-1RAcPL Interleukin-1 receptor 7 IL-1R-7 IL-1R7 Interleukin-18 receptor accessory protein-like Interleukin-18 receptor beta IL-18R-beta IL-18Rbeta CD218b IL18RAP IL1R7; Interleukin-18 receptor accessory protein-like; Interleukin-18 receptor beta
Gene Aliases: ACPL; CD218b; CDw218b; IL-18R-beta; IL-18RAcP; IL-18Rbeta; IL-1R-7; IL-1R7; IL-1RAcPL; IL18RAP; IL18RB; IL1R7
UniProt ID: (Human) O95256, (Mouse) Q9Z2B1
Entrez Gene ID: (Human) 8807, (Rat) 373540, (Mouse) 16174
Molecular Function: cytokine receptor receptor type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.