Invitrogen
Manufacturer Code:MA524806
Catalog # PIMA524806
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 46N7E3 |
Immunogen | Recombit human IL17E which corresponds to Amino acids (97-199). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: IL-17 receptor E; IL-17 receptor E interleukin 17 receptor E; Interleukin-17 receptor E
Gene Aliases: IL17RE; UNQ3056/PRO9877
UniProt ID: (Human) Q8NFR9
Entrez Gene ID: (Human) 132014
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.