Invitrogen
Manufacturer Code:PA534499
Catalog # PIPA534499
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 16 amino acid peptide near the center of human IL-17RE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IL-17 receptor E; IL-17RE; IL-17RE Interleukin-17 receptor E IL-17 receptor E; Interleukin-17 receptor E
Gene Aliases: AA589509; IL-17RE; IL17RE; Il25r; UNQ3056/PRO9877
UniProt ID: (Human) Q8NFR9, (Mouse) Q8BH06, (Rat) Q6AZ51
Entrez Gene ID: (Human) 132014, (Mouse) 57890, (Rat) 362417
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.