Invitrogen
Manufacturer Code:PA521682
Catalog # PIPA521682
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 286 and 531 of IL17 receptor D (Uniprot IDNo.Q8NFM7) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hSef; IL-17 receptor D; IL-17RD; IL17Rhom; Interleukin-17 receptor D; Interleukin-17 receptor-like protein; mSef; Sef homolog; UNQ6115/PRO20026 IL-17RD IL17RLM SEF IL-17 receptor D IL17Rhom hSef interleukin-17 receptor D interleukin-17 receptor-like protein sef homolog
Gene Aliases: 2810004A10Rik; AI428510; HH18; IL-17RD; IL17RD; IL17RLM; SEF; Sef-S; UNQ6115/PRO20026
UniProt ID: (Human) Q8NFM7, (Mouse) Q8JZL1
Entrez Gene ID: (Human) 54756, (Mouse) 171463
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.