Invitrogen
Manufacturer Code:PA547286
Catalog # PIPA547286
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse IL-17 RC Leu21-Trp465 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: IL-17 receptor C; IL-17RL; Interleukin-17 receptor C; Interleukin-17 receptor-like protein; UNQ6118/PRO20040/PRO38901 IL17-RL IL17RL IL-17 receptor C IL-17RL IL17F receptor IL17Rhom interleukin-17 receptor C interleukin-17 receptor homolog interleukin-17 receptor-like protein zcytoR14; ZcytoR14
Gene Aliases: 1110025H02Rik; Gm19850; IL-17RC; IL17-RC; IL17-RL; Il17rc; Il17rl
UniProt ID: (Mouse) Q8K4C2
Entrez Gene ID: (Mouse) 171095
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.