Invitrogen
Manufacturer Code:PA5101326
Catalog # PIPA5101326
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human IL17RA corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AW538159 CANDF5 CD127 CD217 CDw217 hIL-17R IL-17 receptor A Il17r Il17ra IL-17RA interleukin 17 receptor A interleukin-17 receptor A MGC10262 VDw217; CD217; CDw217; IL-17 receptor A; IL-17RA; Interleukin-17 receptor A
Gene Aliases: AW538159; CANDF5; CD217; CDw217; hIL-17R; IL-17RA; IL17R; IL17RA; VDw217
UniProt ID: (Human) Q96F46, (Mouse) Q60943
Entrez Gene ID: (Human) 23765, (Mouse) 16172
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.