Abnova
Manufacturer Code:PAB27447
Catalog # 89-128-958
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 18 amino acids at N-terminus of human IL17RA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of IL17RA.
For Research Use Only
Protein Aliases: CD217; CDw217; hIL-17R; IL-17 receptor A; IL-17RA; IL17R; Interleukin-17 receptor A; MGC10262
Gene Aliases: CANDF5; CD217; CDw217; hIL-17R; IL-17RA; IL17R; IL17RA
UniProt ID: (Human) Q96F46
Entrez Gene ID: (Human) 23765
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.