Abnova
Manufacturer Code:PAB17043
Catalog # 89-118-832
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to mouse Il17ra. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of Il17ra.
For Research Use Only
Protein Aliases: AW538159; CD217; Cdw217; IL-17 receptor A; IL-17RA; Il17r; Interleukin-17 receptor A; VDw217
Gene Aliases: AW538159; Cdw217; Il17r; Il17ra; VDw217
UniProt ID: (Mouse) Q60943
Entrez Gene ID: (Mouse) 16172
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.