Abnova
Manufacturer Code:H00003598M01J
Catalog # 89-335-082
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2E10 |
Immunogen | IL13RA2 (NP_000631 27 a.a. &sim 126 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse Monoclonal Antibody.
Protein Aliases: cancer/testis antigen 19; CD213a2; CD213A2 IL-13R IL13BP; IL-13 receptor subunit alpha-2; IL-13R subunit alpha-2; IL-13R-alpha-2; IL-13RA2; interleukin 13 binding protein; interleukin 13 receptor alpha 2 chain; interleukin 13 receptor, alpha 2; Interleukin-13 receptor subunit alpha-2; Interleukin-13-binding protein
Gene Aliases: CD213A2; CT19; IL-13R; IL13BP; IL13R; IL13RA2
UniProt ID: (Human) Q14627
Entrez Gene ID: (Human) 3598
Molecular Function: cytokine defense/immunity protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.