Invitrogen
Manufacturer Code:PA544066
Catalog # PIPA544066
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human IL13RA2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cancer/testis antigen 19; CD213a2; IL-13 receptor subunit alpha-2; IL-13R subunit alpha-2; IL-13R-alpha-2; IL-13RA2; interleukin 13 binding protein; Interleukin 13 receptor alpha 2 CD213A2 IL-13R IL13BP CT19; interleukin 13 receptor alpha 2 chain; interleukin 13 receptor alpha chain 2; interleukin 13 receptor, alpha 2; Interleukin-13 receptor subunit alpha-2; interleukin-13 receptor subunit alpha-2-like; Interleukin-13-binding protein
Gene Aliases: CD213A2; CT19; IL-13R; IL-13R-alpha-2; IL13BP; IL13R; IL13RA2
UniProt ID: (Dog) Q95LF0, (Human) Q14627, (Mouse) O88786
Entrez Gene ID: (Rabbit) 100348025, (Bovine) 616008, (Dog) 403622, (Pig) 100156991, (Horse) 100057673, (Human) 3598, (Guinea pig) 100733400, (Mouse) 16165, (Rat) 171060
Molecular Function:
cytokine
defense/immunity protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.