OriGene
Manufacturer Code:TA808195
Catalog # 50-168-1962
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A6 |
Immunogen | Human recombit protein fragment corresponding to amino acids 33-283 of human IL12RB1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD212; CD212 IL-12R-BETA1 IL12RB IMD30; cluster of differentiation 212; IL-12 receptor beta component; IL-12 receptor subunit beta-1; IL-12R subunit beta-1; interleukin 12 receptor, beta 1; interleukin-12 receptor beta-1 chain; Interleukin-12 receptor subunit beta-1
Gene Aliases: CD212; IL-12R-BETA1; IL12R; IL12RB; IL12RB1; IMD30
UniProt ID: (Human) P42701
Entrez Gene ID: (Human) 3594
Molecular Function: cytokine defense/immunity protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.