Abnova
Manufacturer Code:MAB16025
Catalog # 89-998-817
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | I 1108 |
Immunogen | Recombinant protein corresponding to human IL12B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CLMF; CLMF p40; CLMF2; Cytotoxic lymphocyte maturation factor 40 kDa subunit; IL-12 subunit p40; IL-12B; IL12, subunit p40; interleukin 12, p40; interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40); interleukin-12 beta chain; Interleukin-12 subunit beta; natural killer cell stimulatory factor, 40 kD subunit; NK cell stimulatory factor chain 2; NKSF; NKSF2
Gene Aliases: CLMF; CLMF2; IL-12B; IL12B; IMD28; IMD29; NKSF; NKSF2
UniProt ID: (Human) P29460
Entrez Gene ID: (Human) 3593
Molecular Function: cytokine interleukin superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.