Novus Biologicals
Manufacturer Code:NBP290961B
Catalog # NB080450
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 68 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human IL12A/IL12B (NP_000873.2 & NP_002178.2 Met1-Ser219 & Met1-Ser328). |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CLMF CLMF p35 CLMF p40 CLMF2 cytotoxic lymphocyte maturation factor 1 p35 Cytotoxic lymphocyte maturation factor 35 kDa subunit Cytotoxic lymphocyte maturation factor 40 kDa subunit IL-12 subunit p35 IL-12A IL-12B IL35 subunit IMD28 IMD29 interleukin 12 p35 interleukin 12 p40 interleukin 12A (natural killer cell stimulatory factor 1 cytotoxic lymphocyte maturation factor 1 p35) interleukin 12B (natural killer cell stimulatory factor 2 cytotoxic lymphocyte maturation factor 2 p40) interleukin-12 alpha chain interleukin-12 beta chain natural killer cell stimulatory factor 40 kD subunit NF cell stimulatory factor chain 1 NFSK NK cell stimulatory factor chain 1 NK cell stimulatory factor chain 2 NKSF NKSF1 NKSF2 p35; CLMF p35; cytotoxic lymphocyte maturation factor 1, p35; Cytotoxic lymphocyte maturation factor 35 kDa subunit; IL-12 subunit p35; IL-12, subunit p35; IL-12A; IL35 subunit; interleukin 12, p35; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); interleukin-12 alpha chain; Interleukin-12 subunit alpha; NF cell stimulatory factor chain 1; NK cell stimulatory factor chain 1; NKSF1
Gene Aliases: CLMF; IL-12A; IL12A; NFSK; NKSF1; P35
UniProt ID: (Human) P29459
Entrez Gene ID: (Human) 3592
Molecular Function: cytokine interleukin superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.