Proteintech
Manufacturer Code:133561AP150UL
Catalog # 50-172-8675
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | IL10RA Fusion Protein Ag4156 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD210; CDw210a; CDW210A HIL 10R IL 10 receptor subunit alpha IL 10R subunit 1 IL 10R subunit alpha IL 10R1 IL 10RA IL-10 R alpha IL10R IL10RA interleukin 10 receptor alpha; IL-10 receptor subunit alpha; IL-10R subunit 1; IL-10R subunit alpha; IL-10RA; interleukin 10 receptor, alpha; interleukin-10 receptor alpha chain; Interleukin-10 receptor subunit 1; Interleukin-10 receptor subunit alpha
Gene Aliases: CD210; CD210a; CDW210A; HIL-10R; IL-10R1; IL10R; IL10RA
UniProt ID: (Human) Q13651
Entrez Gene ID: (Human) 3587
Molecular Function: cytokine receptor defense/immunity protein receptor type I cytokine receptor type II cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.