Invitrogen
Manufacturer Code:MA529850
Catalog # PIMA529850
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 9 |
Immunogen | Recombit Rhesus IL-8/Interleukin-8/CXCL8 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: C-X-C motif chemokine 8; Chemokine (C-X-C motif) ligand 8; IL-8; Interleukin-8
Gene Aliases: CXCL8; IL8
UniProt ID: (Rhesus monkey) P67813
Entrez Gene ID: (Rhesus monkey) 613028
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.