Invitrogen
Manufacturer Code:MA182421
Catalog # PIMA182421
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 8M6 |
Immunogen | Recombit ovine IL-8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4°C or -20°C if preferred |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Alveolar macrophage chemotactic factor I; alveolar macrophage-derived chemotactic factor-I; AMCF-I; C-X-C motif chemokine 8; Chemokine (C-X-C motif) ligand 8; cytokine 8; IL-8; Interleukin-8; NAP-1; neutrophil activating peptide 1; neutrophil and T-lymphocyte chemotactic and activating factor; Neutrophil attractant/activation protein 1; neutrophil attractant/activation protein-1; Permeability factor 1; PF1; RPF1
Gene Aliases: AMCF-I; CXCL8; IL-8; IL8
UniProt ID: (Dog) P41324, (Bovine) P79255, (Sheep) P36925, (Pig) P26894, (Rabbit) P19874
Entrez Gene ID: (Dog) 403850, (Bovine) 280828, (Sheep) 443418, (Pig) 396880, (Rabbit) 100009129
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.