Invitrogen
Manufacturer Code:701276
Catalog # PI701276
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Pig |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 19H2L2 |
Immunogen | Recombit protein corresponding to amino acids 26103 of swine Interleukin 8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Alveolar macrophage chemotactic factor I; alveolar macrophage-derived chemotactic factor-I; AMCF-I; C-X-C motif chemokine 8; Chemokine (C-X-C motif) ligand 8; IL-8; Interleukin-8; Interleukin-8 Monocyte-derived neutrophil chemotactic factor T-cell chemotactic factor Neutrophil-activating protein 1 CXCL8 GCP-1 GCP1 LECT LUCT LYNAP MDNCF MONAP NAF NAP-1 NAP1
Gene Aliases: AMCF-I; CXCL8; IL8
UniProt ID: (Pig) P26894
Entrez Gene ID: (Pig) 396880
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.