Novus Biologicals
Manufacturer Code:NBP225249
Catalog # NBP225249
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 73N8F2 |
Immunogen | Amino acids 145-235 of mouse IL-7R CD127 were used as the immunogen for the antibody. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD127; CD127 antigen; CD127 antigen CD127ILRA CDW127 IL-7 receptor subunit alpha IL-7R subunit alpha IL7RA IL-7RA IL-7R-alpha interleukin 7 receptor interleukin 7 receptor alpha chain interleukin 7 receptor isoform H5-6 interleukin-7 receptor subunit alpha; CDw127; IL-7 receptor subunit alpha; IL-7R subunit alpha; interleukin 7 receptor alpha chain; interleukin 7 receptor isoform H5-6; Interleukin-7 receptor subunit alpha
Gene Aliases: CD127; CDW127; IL-7R-alpha; IL7R; IL7RA; ILRA
UniProt ID: (Human) Q6NWM0
Entrez Gene ID: (Human) 3575
Molecular Function:
cytokine receptor
receptor
type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.