R&D Systems
Manufacturer Code:MAB227100
Catalog # MAB227100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 17506 |
Immunogen | S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-6R alpha Leu20-Asp339 Accession # P08887 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD126; CD126 antigen; CD126 gp80 IL-6 R alpha IL6Q IL6R IL6R alpha IL-6R-1 IL6RA IL-6RA IL6RQ interleukin 6 receptor; gp80; IL-6 receptor subunit alpha; IL-6R 1; Interleukin-6 receptor subunit alpha; Membrane glycoprotein 80; sIL6R; Soluble interleukin-6 receptor subunit alpha
Gene Aliases: CD126; gp80; IL-6R-1; IL-6RA; IL6Q; IL6R; IL6RA; IL6RQ
UniProt ID: (Human) P08887
Entrez Gene ID: (Human) 3570
Molecular Function:
cytokine
defense/immunity protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.