Proteintech
Manufacturer Code:234571AP150UL
Catalog # 50-173-4082
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | IL-6R Fusion Protein Ag18263 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD126; CD126 antigen; CD126 gp80 IL 6 receptor subunit alpha IL 6R 1 IL 6R alpha IL 6R subunit alpha IL 6RA IL-6 R alpha IL-6R IL6RA interleukin 6 receptor Membrane glycoprotein 80; gp80; IL-6 receptor alpha chain; IL-6 receptor subunit alpha; IL-6R 1; IL-6R subunit alpha; IL-6R-alpha; IL-6RA; interleukin 6 receptor, alpha; Interleukin-6 receptor subunit alpha; Membrane glycoprotein 80; sIL6R; Soluble interleukin-6 receptor subunit alpha
Gene Aliases: CD126; gp80; IL-6R; IL-6R-1; IL-6RA; IL6Q; IL6R; IL6R1; IL6RA; IL6RQ
UniProt ID: (Human) P08887, (Mouse) P22272
Entrez Gene ID: (Human) 3570, (Rat) 24499, (Mouse) 16194
Molecular Function:
cytokine
defense/immunity protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.