R&D Systems
Manufacturer Code:MAB2061RSP
Catalog # MAB2061RSP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1936R |
Immunogen | <em>E. coli</em>-derived recombinant human IL-6 Val30-Met212 Accession # P05231 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: B-cell differentiation factor; B-cell differentiation factor B-cell stimulatory factor 2 BSF2 BSF-2 CDF CTL differentiation factor HGF HSF hybridoma growth factor IFNB2 IFN-beta-2 IL6 IL-6 Interferon beta-2 interleukin 6 (interferon beta 2) interleukin BSF-2 interleukin-6 MGI-2A; B-cell stimulatory factor 2; BSF-2; CDF; CTL differentiation factor; Hybridoma growth factor; IFN-beta-2; IL-6; Interferon beta-2; interleukin BSF-2; Interleukin-6
Gene Aliases: BSF-2; BSF2; CDF; HGF; HSF; IFN-beta-2; IFNB2; IL-6; IL6
UniProt ID: (Human) P05231
Entrez Gene ID: (Human) 3569
Molecular Function: cytokine interleukin superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.