R&D Systems
Manufacturer Code:MAB705100
Catalog # MAB705100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | TRFK4 |
Immunogen | Mouse T cell clone-derived partially purified recombinant mouse IL-5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: B-cell differentiation factor I; BCDF mu B-cell differentiation factor I BCGFII EDF Eo-CSF Eosinophil differentiation factor IL5 IL-5T-cell replacing factor interleukin 5 (colony-stimulating factor eosinophil) interleukin-5 TRF TRFB cell differentiation factor I; colony-stimulating factor, eosinophil; Eosinophil differentiation factor; IL-5; Interleukin-5; T-cell replacing factor; TRF
Gene Aliases: EDF; IL-5; IL5; TRF
UniProt ID: (Human) P05113
Entrez Gene ID: (Human) 3567
Molecular Function: cytokine interleukin superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.