R&D Systems
Manufacturer Code:MAB605SP
Catalog # MAB605SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 9906 |
Immunogen | <EM>S. frugiperda</EM> insect ovarian cell line <EM>Sf</EM> 21-derived recombinant human IL-5 Ile20-Ser134 Accession # P05113 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: B-cell differentiation factor I; BCDF mu B-cell differentiation factor I BCGFII EDF Eo-CSF Eosinophil differentiation factor IL5 IL-5T-cell replacing factor interleukin 5 (colony-stimulating factor eosinophil) interleukin-5 TRF TRFB cell differentiation factor I; colony-stimulating factor, eosinophil; Eosinophil differentiation factor; IL-5; Interleukin-5; T-cell replacing factor; TRF
Gene Aliases: EDF; IL-5; IL5; TRF
UniProt ID: (Human) P05113
Entrez Gene ID: (Human) 3567
Molecular Function: cytokine interleukin superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.