Novus Biologicals
Manufacturer Code:NBP288929
Catalog # NB036518
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL9071 |
Immunogen | This antibody was developed against a synthetic peptide corresponding to amino acids: SFRRPFWREEHIEGGHSNTDRPSR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°:C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Fig-1 protein; FIG1 Fig-1 protein hFIG1 IL4-induced protein 1 interleukin 4 induced 1 interleukin four induced 1 LAAO L-amino-acid oxidase LAO UNQ636/PRO1265; hFIG1; IL4-induced protein 1; interleukin four induced 1; Interleukin-4-induced protein 1; L-amino-acid oxidase; LAAO; Protein Fig-1
Gene Aliases: FIG1; IL4I1; LAAO; LAO; UNQ636/PRO1265
UniProt ID: (Human) Q96RQ9
Entrez Gene ID: (Human) 259307
Molecular Function:
DNA binding protein
DNA methyltransferase
methyltransferase
nucleic acid binding
oxidase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.