R&D Systems
Manufacturer Code:MAB5684100
Catalog # MAB5684100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 1006202 |
Immunogen | Chinese Hamster Ovary cell line CHO-derived human IL-4I1 Met1-His567 Accession No. Q96RQ9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Rat Monoclonal Antibody
Protein Aliases: Fig-1 protein; FIG1 Fig-1 protein hFIG1 IL4I1 IL4-induced protein 1 interleukin 4 induced 1 interleukin four induced 1 LAAO L-amino-acid oxidase LAO UNQ636/PRO1265; hFIG1; IL4-induced protein 1; interleukin four induced 1; Interleukin-4-induced protein 1; L-amino-acid oxidase; LAAO; Protein Fig-1
Gene Aliases: FIG1; IL4I1; LAAO; LAO; UNQ636/PRO1265
UniProt ID: (Human) Q96RQ9
Entrez Gene ID: (Human) 259307
Molecular Function:
DNA binding protein
DNA methyltransferase
methyltransferase
nucleic acid binding
oxidase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.