R&D Systems
Manufacturer Code:MAB304SP
Catalog # MAB304SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3007 |
Immunogen | <EM>E. coli</EM>-derived recombinant human IL-4 His25-Ser153 Accession # P05112.1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: B cell growth factor 1; B cell growth factor 1 BCDF B-cell stimulatory factor 1 BCGF1 BCGF-1 binetrakin BSF1 BSF-1 IL4 IL-4B_cell stimulatory factor 1 IL4E12 interleukin 4 interleukin-4 Lymphocyte stimulatory factor 1 MGC79402 Pitrakinra; B-cell stimulatory factor 1; B_cell stimulatory factor 1; Binetrakin; BSF-1; IL-4; interleukin 4 variant 2; Interleukin-4; Lymphocyte stimulatory factor 1; Pitrakinra
Gene Aliases: BCGF-1; BCGF1; BSF-1; BSF1; IL-4; IL4
UniProt ID: (Human) P05112
Entrez Gene ID: (Human) 3565
Molecular Function: cytokine interleukin superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.