Invitrogen
Manufacturer Code:M450B
Catalog # ENM450B
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 25D2 |
Immunogen | Recombit human IL-4 (CHO cell-derived) |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: B cell growth factor 1; B cell growth factor 1 BSF-1 B&lowbarcell stimulatory factor 1 binetrakin interleukin-4 lymphocyte stimulatory factor 1 pitrakinra; B-cell stimulatory factor 1; B_cell stimulatory factor 1; Binetrakin; BSF-1; IL-4; interleukin 4 variant 2; Interleukin-4; Lymphocyte stimulatory factor 1; Pitrakinra
Gene Aliases: BCGF-1; BCGF1; BSF-1; BSF1; IL-4; IL4
UniProt ID: (Human) P05112
Entrez Gene ID: (Human) 3565
Molecular Function: cytokine interleukin superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.