Invitrogen
Manufacturer Code:PA547930
Catalog # PIPA547930
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit canine IL-4 His25-His132 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: B cell growth factor 1 BSF-1 B&lowbarcell stimulatory factor 1 binetrakin interleukin-4 lymphocyte stimulatory factor 1 pitrakinra; B-cell stimulatory factor 1; BSF-1; cytokine; IL-4; Interleukin-4; Lymphocyte stimulatory factor 1
Gene Aliases: IL-4; IL4
UniProt ID: (Dog) O77762
Entrez Gene ID: (Dog) 403785
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.