Novus Biologicals
Manufacturer Code:NBP289550100UL
Catalog # NB020583
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 17 |
Immunogen | Recombinant Human IL-3R alpha/CD123 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CD123; CD123 antigen; CD123 CD123 antigen hIL3Ra hIL-3Ra IL-3 receptor subunit alpha IL3R IL-3R subunit alpha IL-3RA IL-3R-alpha IL3RAY IL3RX IL3RY interleukin 3 receptor alpha (low affinity) interleukin-3 receptor subunit alpha MGC34174; IL-3 receptor alpha SP2 isoform; IL-3 receptor subunit alpha; IL-3R subunit alpha; IL-3R-alpha; IL-3RA; interleukin 3 receptor, alpha (low affinity); Interleukin-3 receptor subunit alpha
Gene Aliases: CD123; hIL-3Ra; IL3R; IL3RA; IL3RAY; IL3RX; IL3RY
UniProt ID: (Human) P26951
Entrez Gene ID: (Human) 3563
Molecular Function: cytokine defense/immunity protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.