Novus Biologicals
Manufacturer Code:NBP176356
Catalog # NBP176356
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 13 amino acid synthetic peptide from near the center of human IL-34. The immunogen is located within amino acids 60 - 110 of IL-34. Amino Acid Squence: TRLRAQVSERELR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C16orf77 IL-34 interleukin 34; IL-34; Interleukin-34
Gene Aliases: C16orf77; IL-34; IL34
UniProt ID: (Human) Q6ZMJ4
Entrez Gene ID: (Human) 146433
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.