Invitrogen
Manufacturer Code:25933580
Catalog # 50-112-3451
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | RMST2-2 |
Conjugate | PE-Cyanine7 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: DER4 Fit-1 Ly84 ST2L St2 St2-rs1 T1 T1/ST2; Interleukin-1 receptor-like 1; Interleukin-33 receptor alpha chain; Lymphocyte antigen 84; Protein ST2; Protein T1
Gene Aliases: DER4; Fit-1; Il1rl1; Ly84; St2; St2-rs1; ST2L; Ste2; T1; T1/ST2
UniProt ID: (Mouse) P14719
Entrez Gene ID: (Mouse) 17082
Molecular Function: cytokine receptor receptor type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.