Novus Biologicals
Manufacturer Code:NBP175516
Catalog # NBP175516
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 12B3C4 |
Immunogen | Antibody was raised against the recombinant IL-33 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20 degrees C. Avoid freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C9orf26chromosome 9 open reading frame 26 (NF-HEV) DKFZp586H0523 DVS27 DVS27-related protein IL1F11 IL-1F11 IL-33 interleukin 33 Interleukin-1 family member 11 interleukin-33 NFHEV NF-HEVNFEHEV Nuclear factor from high endothelial venules RP11-575C20.2; DVS27-related protein; IL-1F11; IL-33; Interleukin-1 family member 11; Interleukin-33; Interleukin-33 (109-270); Interleukin-33 (95-270); Interleukin-33 (99-270); NF-HEV; nuclear factor for high endothelial venules; Nuclear factor from high endothelial venules
Gene Aliases: C9orf26; DVS27; IL1F11; IL33; NF-HEV; NFEHEV; NFHEV
UniProt ID: (Human) O95760
Entrez Gene ID: (Human) 90865
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.