Invitrogen
Manufacturer Code:PA579499
Catalog # PIPA579499
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived rat IL33 recombit protein (Position: A102-M264). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IL-33; Interleukin-33; Interleukin-33 IL-33 Il33; Interleukin-33(102-266); Interleukin-33(109-266); nuclear factor from high endothelial venules
Gene Aliases: 9230117N10Rik; Il-33; Il1f11; Il33; NF-HEV; RGD1311155
UniProt ID: (Mouse) Q8BVZ5, (Rat) Q66H70
Entrez Gene ID: (Mouse) 77125, (Rat) 361749
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.