Invitrogen
Manufacturer Code:PA547007
Catalog # PIPA547007
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli derived recombit mouse IL 33 Ser109-Ile266 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: C9orf26 DVS27 IL1F11 NF-HEV NFEHEV RP11-575C20.2 DVS27-related protein IL-1F11 interleukin-1 family member 11 interleukin-33 nuclear factor for high endothelial venules nuclear factor from high endothelial venules; IL-33; Interleukin-33; Interleukin-33(102-266); Interleukin-33(109-266); nuclear factor from high endothelial venules
Gene Aliases: 9230117N10Rik; Il-33; Il1f11; Il33; NF-HEV
UniProt ID: (Mouse) Q8BVZ5
Entrez Gene ID: (Mouse) 77125
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.