Invitrogen
Manufacturer Code:PA520398
Catalog # PIPA520398
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A peptide from near the center of human IL-33 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DVS27-related protein; IL-1F11; IL-33; Interleukin-1 family member 11; Interleukin-33; Interleukin-33 (109-270); Interleukin-33 (95-270); Interleukin-33 (99-270); Interleukin-33(102-266); Interleukin-33(109-266); NF-HEV; nuclear factor for high endothelial venules; Nuclear factor from high endothelial venules
Gene Aliases: 9230117N10Rik; C9orf26; DVS27; Il-33; IL1F11; IL33; NF-HEV; NFEHEV; NFHEV
UniProt ID: (Human) O95760, (Mouse) Q8BVZ5
Entrez Gene ID: (Human) 90865, (Mouse) 77125
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.