OriGene
Manufacturer Code:TA500182S
Catalog # 50-167-1418
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3B11 |
Immunogen | Human recombit protein fragment corresponding to amino acids 20-152 of human IL-3 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: colony-stimulating factor, multiple; Hematopoietic growth factor; IL-3; IL-3 MCGF MULTI-CSF; Interleukin-3; Mast cell growth factor; mast-cell growth factor; MCGF; multilineage-colony-stimulating factor; Multipotential colony-stimulating factor; P-cell stimulating factor; P-cell-stimulating factor
Gene Aliases: IL-3; IL3; MCGF; MULTI-CSF
UniProt ID: (Human) P08700
Entrez Gene ID: (Human) 3562
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.