Novus Biologicals
Manufacturer Code:NBP2117070.025MG
Catalog # NB018864
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Blocking Assay (BLOCK) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0401-10X23 |
Immunogen | Human IL29 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cytokine Zcyto21; cytokine Zcyto21 IFNL1 IFN-lambda-1 IL-29 interferon lambda-1 interferon lambda 1 interleukin 29 (interferon lambda 1) interleukin-29 ZCYTO21; IFN-lambda-1; IL-29; Interferon lambda-1; interferon, lambda 1; interferon-lambda-1; interleukin 29 (interferon, lambda 1); Interleukin-29
Gene Aliases: IFNL1; IL-29; IL29; ZCYTO21
UniProt ID: (Human) Q8IU54
Entrez Gene ID: (Human) 282618
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.