R&D Systems
Manufacturer Code:AF1598
Catalog # AF1598
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human IL-29/IFN-lambda 1 Gly20-Thr200 Accession # Q8IU54 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Cytokine Zcyto21; cytokine Zcyto21 IFNL1 IFN-lambda 1 IFN-lambda-1 IL29 IL-29 interferon lambda-1 interferon lambda 1 interleukin 29 (interferon lambda 1) interleukin-29 ZCYTO21; IFN-lambda-1; IL-29; Interferon lambda-1; interferon, lambda 1; interferon-lambda-1; interleukin 29 (interferon, lambda 1); Interleukin-29
Gene Aliases: IFNL1; IL-29; IL29; ZCYTO21
UniProt ID: (Human) Q8IU54
Entrez Gene ID: (Human) 282618
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.