Invitrogen
Manufacturer Code:MA530680
Catalog # PIMA530680
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3 |
Immunogen | Recombit Human IL-29 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cytokine Zcyto21; IFN-lambda-1; IL-29; Interferon lambda-1; interferon, lambda 1; interferon-lambda-1; interleukin 29 (interferon, lambda 1); Interleukin-29; Interleukin-29 IL-29 Interferon lambda-1 IFN-lambda-1 Cytokine ZCYTO21 FNL1 ZCYTO21
Gene Aliases: IFNL1; IL-29; IL29; ZCYTO21
UniProt ID: (Human) Q8IU54
Entrez Gene ID: (Human) 282618
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.