Invitrogen
Manufacturer Code:MA523903
Catalog # PIMA523903
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 247801 |
Immunogen | Mouse myeloma cell line NS0-derived recombit human IL-29/IFN-lambda 1 Gly20-Thr200 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cytokine Zcyto22; IFN-lambda-3; IFN-lambda-4; IL-28B; IL-28C; Interferon lambda-3; interferon, lambda 3; interferon, lambda 4; Interleukin-28B; Interleukin-28C; Interleukin-29 IL-29 Interferon lambda-1 IFN-lambda-1 Cytokine ZCYTO21 FNL1 ZCYTO21
Gene Aliases: IFNL3; IL-28B; IL28B; IL28C; ZCYTO22
UniProt ID: (Human) Q8IZI9
Entrez Gene ID: (Human) 282617
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.