R&D Systems
Manufacturer Code:MAB15874SP
Catalog # MAB15874SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 884136R |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human IL-28A/IFN-lambda 2 Val26-Val200 Accession # NP_742150 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
Protein Aliases: Cytokine Zcyto20; IFN-lambda-2; IL-28A; Interferon lambda-2; interferon, lambda 2; interferon-lambda 2; interleukin 28A (interferon, lambda 2); Interleukin-28A; interleukin-28A IFN-lambda 2 IL28A IL-28A interferon lambda 2
Gene Aliases: IFNL2; IL-28A; IL28A; ZCYTO20
UniProt ID: (Human) Q8IZJ0
Entrez Gene ID: (Human) 282616
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.