R&D Systems
Manufacturer Code:FAB14791G025
Catalog # FAB14791G02
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 191106 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human IL-27 R alpha/WSX-1/TCCR Gly34-Lys516 Accession # Q6UWB1 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: class I cytokine receptor; class I cytokine receptor CRL1IL-27R Cytokine receptor-like 1 IL-27 R alpha IL27R IL27R alpha IL27RA IL-27RA IL-27R-alpha interleukin 27 receptor alpha interleukin-27 receptor subunit alpha TCCR TCCRIL-27 receptor subunit alpha T-cell cytokine receptor type 1 Type I T-cell cytokine receptor WSX-1 WSX1IL-27R subunit alpha zcytor1; Cytokine receptor WSX-1; Cytokine receptor-like 1; IL-27 receptor subunit alpha; IL-27R subunit alpha; IL-27R-alpha; interleukin 27 receptor, alpha; Interleukin-27 receptor subunit alpha; T-cell cytokine receptor type 1; TCCR; Type I T-cell cytokine receptor; ZcytoR1
Gene Aliases: CRL1; IL-27RA; IL27R; IL27RA; TCCR; UNQ296/PRO336; WSX1; zcytor1
UniProt ID: (Human) Q6UWB1
Entrez Gene ID: (Human) 9466
Molecular Function:
cytokine
defense/immunity protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.