Novus Biologicals
Manufacturer Code:NBP176728SS
Catalog # NBP176728SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a synthetic peptide corresponding to amino acids near the amino terminus of human TCCR precursor. The immunogen is located within the first 50 amino acids of TCCR. Amino Acid Squence: GSAGPLQCYGVGPLGD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
Protein Aliases: class I cytokine receptor; class I cytokine receptor CRL1IL-27R Cytokine receptor-like 1 IL27R IL-27RA IL-27R-alpha interleukin 27 receptor alpha interleukin-27 receptor subunit alpha TCCRIL-27 receptor subunit alpha T-cell cytokine receptor type 1 Type I T-cell cytokine receptor WSX-1 WSX1IL-27R subunit alpha zcytor1; Cytokine receptor WSX-1; Cytokine receptor-like 1; IL-27 receptor subunit alpha; IL-27R subunit alpha; IL-27R-alpha; interleukin 27 receptor, alpha; Interleukin-27 receptor subunit alpha; T-cell cytokine receptor type 1; TCCR; Type I T-cell cytokine receptor; ZcytoR1
Gene Aliases: CRL1; IL-27RA; IL27R; IL27RA; TCCR; UNQ296/PRO336; WSX1; zcytor1
UniProt ID: (Human) Q6UWB1
Entrez Gene ID: (Human) 9466
Molecular Function:
cytokine
defense/immunity protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.