Novus Biologicals
Manufacturer Code:NBP211702UV
Catalog # NBP211702UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0398-6F48 |
Immunogen | Human Interleukin-24 |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: C49A FISP IL10B IL-24 interleukin 24 mda-7 MDA7MOB5 melanocyte-associated Mda-7 melanoma differentiation association protein 7 Melanoma differentiation-associated gene 7 protein Mob-5 ST16interleukin-24 suppression of tumorigenicity 16 (melanoma differentiation) Suppression of tumorigenicity 16 protein; IL-24; IL-4-induced secreted protein; Interleukin-24; MDA-7; melanocyte-associated Mda-7; Melanoma differentiation-associated gene 7 protein; suppression of tumorigenicity 16 (melanoma differentiation); Suppression of tumorigenicity 16 protein
Gene Aliases: C49A; FISP; IL10B; IL24; MDA7; MOB5; ST16
UniProt ID: (Human) Q13007
Entrez Gene ID: (Human) 11009
Molecular Function: cytokine interleukin superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.