Novus Biologicals
Manufacturer Code:NBP290083AF700
Catalog # NB069666
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 101 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human IL-24 (NP_001172085.1 Met1-Leu207). |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: C49A FISP IL10B IL-24 interleukin 24 mda-7 MDA7MOB5 melanocyte-associated Mda-7 melanoma differentiation association protein 7 Melanoma differentiation-associated gene 7 protein Mob-5 ST16interleukin-24 suppression of tumorigenicity 16 (melanoma differentiation) Suppression of tumorigenicity 16 protein; IL-24; IL-4-induced secreted protein; Interleukin-24; MDA-7; melanocyte-associated Mda-7; Melanoma differentiation-associated gene 7 protein; suppression of tumorigenicity 16 (melanoma differentiation); Suppression of tumorigenicity 16 protein
Gene Aliases: C49A; FISP; IL10B; IL24; MDA7; MOB5; ST16
UniProt ID: (Human) Q13007
Entrez Gene ID: (Human) 11009
Molecular Function: cytokine interleukin superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.