R&D Systems
Manufacturer Code:AF1965SP
Catalog # AF1965SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human IL-24 Gln50-Leu206 Accession # Q2YHE5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: C49A FISP IL10B IL24 IL-24 interleukin 24 mda-7 MDA7MOB5 melanocyte-associated Mda-7 melanoma differentiation association protein 7 Melanoma differentiation-associated gene 7 protein MOB-5 ST16 ST16interleukin-24 suppression of tumorigenicity 16 (melanoma differentiation) Suppression of tumorigenicity 16 protein; IL-24; IL-4-induced secreted protein; Interleukin-24; MDA-7; melanocyte-associated Mda-7; Melanoma differentiation-associated gene 7 protein; suppression of tumorigenicity 16 (melanoma differentiation); Suppression of tumorigenicity 16 protein
Gene Aliases: C49A; FISP; IL10B; IL24; MDA7; MOB5; ST16
UniProt ID: (Human) Q13007
Entrez Gene ID: (Human) 11009
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.